Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183250.2 | NP_899073.1 | 4 | 212 | ||
GenBank mRNA | AK002863.1 | - | 4 | 213 | ||
AK003762.1 | - | 4 | 213 | |||
AK160705.1 | - | 4 | 210 | |||
AK169860.1 | - | 4 | 206 | |||
BC034602.1 | - | 4 | 194 | |||
BC054363.1 | - | 4 | 185 | |||
BC090400.1 | - | 4 | 185 | |||
BC106104.1 | - | 4 | 184 | 5029265 | ||
BC116987.1 | - | 4 | 208 | 40126306 | ||
BC116989.1 | - | 4 | 208 | 40126308 |