Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039647.2 | NP_001034736.2 | 11 | 1893 | ||
GenBank mRNA | BC111039.1 | - | 11 | 1890 | 5349539 |