Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000333.3 | NP_000324.1 | 12 | 2930 | ||
NM_001128149.2 | NP_001121621.2 | 8 | 2110 | |||
NM_001177387.1 | NP_001170858.1 | 11 | 2487 | |||
GenBank mRNA | AF032103.1 | - | 10 | 2069 | ||
AF032105.1 | - | 12 | 2930 | |||
AJ000517.1 | - | 12 | 2938 | |||
AK304062.1 | - | 8 | 2033 |