Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pydc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177349.1 | NP_001170820.1 | 3 | 564 | ||
NM_001177350.1 | NP_001170821.1 | 2 | 512 | |||
GenBank mRNA | AA170499.1 | - | 1 | 46 | ||
AK143015.1 | - | 3 | 565 |
Gene Symbol: Pydc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162938.1 | NP_001156410.1 | 2 | 544 | ||
GenBank mRNA | AK036671.1 | - | 2 | 314 | ||
AK041099.1 | - | 2 | 546 |