Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF508795.1 | - | 6 | 818 | ||
BC062403.1 | - | 6 | 790 | 5621394 | ||
FQ209590.1 | - | 6 | 801 | |||
FQ218571.1 | - | 6 | 803 | |||
FQ218640.1 | - | 1 | 123 | |||
FQ220674.1 | - | 6 | 801 | |||
M15327.1 | - | 6 | 774 |