Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glul | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017073.3 | NP_058769.4 | 4 | 533 | ||
GenBank mRNA | BC061559.1 | - | 4 | 546 | 6918514 | |
BC072694.1 | - | 4 | 533 | 7103110 | ||
FQ213256.1 | - | 4 | 544 | |||
FQ216093.1 | - | 4 | 538 | |||
FQ226911.1 | - | 4 | 556 | |||
FQ227474.1 | - | 4 | 545 | |||
FQ232853.1 | - | 4 | 594 | |||
FQ232963.1 | - | 4 | 545 | |||
FQ233770.1 | - | 4 | 545 | |||
FQ234289.1 | - | 4 | 546 | |||
M29579.1 | - | 4 | 544 | |||
M91652.1 | - | 4 | 542 | |||
X07921.1 | - | 4 | 544 |