Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cth | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017074.1 | NP_058770.1 | 3 | 299 | ||
XM_006233536.2 | XP_006233598.1 | 2 | 238 | |||
GenBank mRNA | AB052882.2 | - | 3 | 298 | ||
AY032875.1 | - | 3 | 298 | |||
AY641456.1 | - | 3 | 298 | |||
BC078869.1 | - | 3 | 312 | 7109746 | ||
D17370.1 | - | 3 | 299 | |||
X53460.1 | - | 187 |
Gene Symbol: LOC103691744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761574.1 | XP_008759796.1 | 346 |