Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000478.5 | NP_000469.3 | 2 | 308 | ||
XM_005245818.1 | XP_005245875.1 | 2 | 278 | |||
XM_006710546.2 | XP_006710609.1 | 2 | 192 | |||
GenBank mRNA | AB011406.1 | - | 2 | 220 | ||
AB209814.1 | - | 2 | 280 | |||
AK293184.1 | - | 2 | 226 | |||
AK312667.1 | - | 2 | 240 | |||
BC021289.2 | - | 2 | 232 | 5087597 | ||
BC066116.1 | - | 2 | 228 | 5754297 | ||
BC090861.1 | - | 2 | 221 | 6181160 | ||
BC110909.1 | - | 2 | 208 | 5752191 | ||
BC126165.1 | - | 1 | 157 | 8991881 | ||
BC136325.1 | - | 1 | 157 | 9020312 | ||
CN425768.1 | - | 2 | 299 | |||
DA574668.1 | - | 2 | 295 | |||
DA625627.1 | - | 2 | 263 | |||
DA631560.1 | - | 2 | 267 | |||
HY052024.1 | - | 2 | 309 |