Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013004.1 | NP_037136.1 | 3 | 752 | ||
GenBank mRNA | AJ001637.1 | - | 3 | 752 |