Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024361.1 | NP_077337.1 | 7 | 1929 | ||
XM_008772130.1 | XP_008770352.1 | 3 | 317 | |||
GenBank mRNA | M92042.1 | - | 7 | 1929 |