Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031604.2 | NP_113792.2 | 5 | 469 | ||
XM_006247295.2 | XP_006247357.1 | 5 | 469 | |||
XM_006247296.2 | XP_006247358.1 | 5 | 469 | |||
XM_006247297.2 | XP_006247359.1 | 5 | 469 | |||
XM_008768002.1 | XP_008766224.1 | 5 | 432 | |||
XM_008768003.1 | XP_008766225.1 | 5 | 432 | |||
XM_008768004.1 | XP_008766226.1 | 5 | 470 | |||
GenBank mRNA | DQ286421.1 | - | 4 | 311 | ||
DQ286422.1 | - | 4 | 311 | |||
DQ286423.1 | - | 4 | 311 | |||
DQ286424.1 | - | 4 | 311 | |||
M58758.1 | - | 4 | 341 |