Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Espn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239476.2 | XP_006239538.1 | 7 | 1421 | ||
XM_006239479.2 | XP_006239541.1 | 7 | 1421 | |||
XM_008764375.1 | XP_008762597.1 | 7 | 1430 | |||
XM_008764376.1 | XP_008762598.1 | 7 | 1430 | |||
XM_008764377.1 | XP_008762599.1 | 7 | 1430 | |||
XM_008764378.1 | XP_008762600.1 | 7 | 1430 | |||
XM_008764379.1 | XP_008762601.1 | 7 | 1430 | |||
XM_008764380.1 | XP_008762602.1 | 7 | 1430 | |||
GenBank mRNA | AF540946.1 | - | 2 | 250 | ||
AF540947.1 | - | 2 | 250 | |||
AF540948.1 | - | 2 | 250 | |||
AF540949.1 | - | 2 | 250 | |||
U46007.1 | - | 7 | 1244 |