Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206847.1 | NP_996729.1 | 8 | 848 | ||
XM_008771885.1 | XP_008770107.1 | 8 | 908 | |||
GenBank mRNA | BC088847.1 | - | 8 | 848 | 7312293 | |
FN432820.1 | - | 8 | 876 | |||
L25387.1 | - | 8 | 822 |