Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130894.4 | NP_570964.4 | 2 | 194 | ||
XM_006239402.2 | XP_006239464.1 | 3 | 347 | |||
XM_006239403.1 | XP_006239465.1 | 3 | 376 | |||
XM_008764288.1 | XP_008762510.1 | 4 | 364 | |||
GenBank mRNA | BC070880.1 | - | 2 | 198 | 7096620 | |
U41803.2 | - | 3 | 212 |
Gene Symbol: LOC100911485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750086.3 | XP_003750134.1 | 667 |