Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005420.2 | NP_005411.1 | 6 | 670 | ||
XM_011532210.2 | XP_011530512.1 | 7 | 697 | |||
GenBank mRNA | AK312236.1 | - | 5 | 569 | ||
BC027956.1 | - | 6 | 670 | 5210178 | ||
CR407621.1 | - | 5 | 557 | |||
S77383.1 | - | 6 | 593 | |||
U08098.1 | - | 6 | 663 | |||
U55764.1 | - | 1 | 16 | |||
Y11195.1 | - | 6 | 619 |