Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031144.3 | NP_112406.1 | 5 | 908 | ||
GenBank mRNA | AB028846.1 | - | 2 | 117 | ||
BC063166.1 | - | 5 | 911 | 6920838 | ||
EF156276.1 | - | 3 | 630 | |||
FQ226945.1 | - | 182 | ||||
FQ227027.1 | - | 470 | ||||
FQ227201.1 | - | 267 | ||||
FQ228822.1 | - | 489 | ||||
FQ229293.1 | - | 269 | ||||
FQ231277.1 | - | 264 | ||||
FQ231503.1 | - | 5 | 910 | |||
FQ231691.1 | - | 258 | ||||
FQ232063.1 | - | 5 | 910 | |||
FQ232650.1 | - | 5 | 910 | |||
FQ232682.1 | - | 450 |