Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170558.1 | NP_001164029.1 | 8 | 9559 | ||
NM_001170559.1 | NP_001164030.1 | 5 | 1453 | |||
NM_001170560.1 | NP_001164031.1 | 6 | 4351 | |||
NM_053663.1 | NP_446115.1 | 8 | 6759 | |||
GenBank mRNA | AF062402.1 | - | 5 | 6380 | ||
AF072892.1 | - | 1453 |