Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oplah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053904.1 | NP_446356.1 | 13, 14 | 1942 | ||
XM_006241771.2 | XP_006241833.1 | 13, 14 | 3602 | |||
XM_006241772.2 | XP_006241834.1 | 13, 14 | 3602 | |||
XM_006241773.2 | XP_006241835.1 | 13, 14 | 3485 | |||
XM_006241774.2 | XP_006241836.1 | 13, 14 | 2280 | |||
XM_006241775.2 | XP_006241837.1 | 14, 15 | 2139 | |||
GenBank mRNA | BC085330.1 | - | 13, 14 | 2007 | 7096390 | |
U70825.1 | - | 13, 14 | 1942 |