Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130421.2 | NP_569105.1 | 4 | 404 | ||
GenBank mRNA | BC100066.1 | - | 4 | 404 | 7387128 | |
FQ232596.1 | - | 4 | 418 |