Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138832.1 | NP_620187.1 | 11 | 1074 | ||
XM_006242277.2 | XP_006242339.1 | 11 | 1133 | |||
XM_006242278.2 | XP_006242340.1 | 12 | 1342 | |||
XM_006242279.2 | XP_006242341.1 | 12 | 1312 | |||
XM_006242280.2 | XP_006242342.1 | 11 | 1181 | |||
XM_006242281.2 | XP_006242343.1 | 11 | 1222 | |||
GenBank mRNA | AF075704.1 | - | 11 | 1074 | ||
BC097283.1 | - | 11 | 994 | 7444889 |