Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcng3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033957.1 | NP_001029129.1 | 2 | 1141 | ||
NM_133426.2 | NP_596917.2 | 2 | 1174 | |||
GenBank mRNA | AB070605.1 | - | 2 | 868 | ||
AF454549.1 | - | 2 | 1325 | |||
AF454550.1 | - | 2 | 1358 |