Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nacc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134413.2 | NP_599240.1 | 6 | 1595 | ||
XM_006255214.2 | XP_006255276.1 | 6 | 1589 | |||
XM_006255215.2 | XP_006255277.1 | 6 | 1509 | |||
XM_008772346.1 | XP_008770568.1 | 6 | 1612 | |||
GenBank mRNA | AF015911.1 | - | 7 | 1596 |