Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000358.2 | NP_000349.1 | 5 | 732 | ||
GenBank mRNA | AB209598.1 | - | 1 | 1218 | ||
AK093916.1 | - | 5 | 587 | |||
AK094055.1 | - | 6 | 663 | |||
AK094581.1 | - | 5 | 676 | |||
AK222833.1 | - | 5 | 640 | |||
AK303643.1 | - | 5 | 964 | |||
BC000097.2 | - | 5 | 625 | 3506051 | ||
BC004972.1 | - | 5 | 620 | 2958878 | ||
BT009820.1 | - | 5 | 571 | |||
KU178357.1 | - | 5 | 571 | |||
KU178358.1 | - | 5 | 571 | |||
M77349.1 | - | 5 | 618 |