Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a6os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139328.2 | NP_647544.2 | 3 | 512 | ||
GenBank mRNA | AY098917.2 | - | 2 | 180 | ||
DY567141.1 | - | 3 | 512 | |||
FQ211988.1 | - | 3 | 519 | |||
FQ220945.1 | - | 3 | 523 | |||
FQ224990.1 | - | 2 | 244 | |||
FQ233662.1 | - | 3 | 521 | |||
FQ234848.1 | - | 3 | 519 |