Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022037.2 | NP_071320.2 | 9 | 882 | ||
NM_022173.2 | NP_071505.2 | 10 | 915 | |||
XM_005264527.1 | XP_005264584.1 | 9 | 892 | |||
XM_005264528.4 | XP_005264585.1 | 9 | 827 | |||
XM_005264531.1 | XP_005264588.1 | 11 | 962 | |||
XM_011533077.1 | XP_011531379.1 | 10 | 925 | |||
XM_011533078.1 | XP_011531380.1 | 9 | 4588 | |||
XM_011533081.2 | XP_011531383.1 | 10 | 1688 | |||
XM_011533082.2 | XP_011531384.1 | 8 | 5260 | |||
XM_017004789.1 | XP_016860278.1 | 10 | 744 | |||
XM_017004790.1 | XP_016860279.1 | 9 | 710 | |||
XM_017004791.1 | XP_016860280.1 | 8 | 794 | |||
XM_017004792.1 | XP_016860281.1 | 9 | 3099 | |||
XM_017004793.1 | XP_016860282.1 | 9 | 2924 | |||
XM_017004794.1 | XP_016860283.1 | 9 | 1656 | |||
XM_017004795.1 | XP_016860284.1 | 11 | 922 | |||
XM_017004796.1 | XP_016860285.1 | 9 | 4430 | |||
XM_017004797.1 | XP_016860286.1 | 10 | 2226 | |||
XM_017004798.1 | XP_016860287.1 | 10 | 957 | |||
XM_017004799.1 | XP_016860288.1 | 10 | 919 | |||
XM_017004800.1 | XP_016860289.1 | 9 | 886 | |||
GenBank mRNA | AB209211.1 | - | 10 | 1059 | ||
DR157578.1 | - | 4 | 254 | |||
M77142.1 | - | 9 | 721 |