Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004442.1 | NP_001004442.1 | 8 | 747 | ||
GenBank mRNA | BC082063.1 | - | 8 | 747 | 7132589 | |
FQ229584.1 | - | 3 | 205 | |||
X79328.1 | - | 7 | 687 |