Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APBB1 SIRNA Assays ›
Gene Symbol:
APBB1
Gene Name:
amyloid beta precursor protein binding family B member 1
Gene Aliases:
FE65, MGC:9072, RIR
Chromosome Location:
Chr.11: 6395124 - 6419830 on Build GRCh38
UniGene ID:
Hs.372840
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s1429
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APBB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164.4
NP_001155.1
12
1796
NM_001257319.2
NP_001244248.1
11
1123
NM_001257320.2
NP_001244249.1
11
1211
NM_001257321.2
NP_001244250.1
12
1259
NM_001257323.2
NP_001244252.1
10
1117
NM_001257325.2
NP_001244254.1
11
1019
NM_001257326.2
NP_001244255.1
11
1219
NM_145689.2
NP_663722.1
11
1790
NR_047512.2
12
1796
XM_006718209.3
XP_006718272.2
11
2134
XM_006718210.3
XP_006718273.2
11
2169
XM_006718211.1
XP_006718274.1
11
2340
XM_006718212.3
XP_006718275.2
12
1846
XM_006718213.1
XP_006718276.1
11
2325
XM_006718214.3
XP_006718277.2
11
1840
XM_006718215.3
XP_006718278.3
12
2154
XM_011520037.2
XP_011518339.1
10
1820
XM_011520038.2
XP_011518340.1
10
1325
XM_011520040.2
XP_011518342.1
11
1034
XM_011520042.1
XP_011518344.1
10
1315
XM_017017639.1
XP_016873128.1
12
2052
XM_017017640.1
XP_016873129.1
11
2046
XM_017017641.1
XP_016873130.1
11
2031
XM_017017642.1
XP_016873131.1
10
1526
GenBank mRNA
AF394214.1
-
12
1755
AK293452.1
-
13
1300
AK293550.1
-
11
1217
AK293554.1
-
11
987
AK293613.1
-
11
851
AK293643.1
-
11
1211
AK295241.1
-
10
1082
AK295425.1
-
11
1008
AK297544.1
-
13
1323
AK297550.1
-
12
1260
AK311141.1
-
11
1052
AK315965.1
-
11
1210
BC010854.2
-
11
1744
3905603
BX538185.1
-
11
1033
EF103274.2
-
11
1067
L77864.1
-
12
1755
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
cellular response to DNA damage stimulus
cell cycle arrest
signal transduction
axonogenesis
response to iron ion
positive regulation of neuron projection development
negative regulation of cell growth
positive regulation of apoptotic process
histone H4 acetylation
positive regulation of DNA repair
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein secretion
negative regulation of thymidylate synthase biosynthetic process
beta-amyloid binding
chromatin binding
protein binding
transcription factor binding
protein complex binding
histone binding
tau protein binding
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-c74kb:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0