Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNS1 SIRNA Assays ›
Gene Symbol:
TNS1
Gene Name:
tensin 1
Gene Aliases:
MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS
Chromosome Location:
Chr.2: 217799789 - 217978853 on Build GRCh38
UniGene ID:
Hs.471381
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14301
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TNS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308022.1
NP_001294951.1
24
4684
NM_001308023.1
NP_001294952.1
23
4660
NM_022648.5
NP_072174.3
26
4932
XM_011511712.1
XP_011510014.1
24
4915
XM_011511713.1
XP_011510015.1
24
4900
XM_011511715.1
XP_011510017.1
23
4876
XM_011511723.2
XP_011510025.1
24
4876
XM_011511728.1
XP_011510030.1
8
5968
XM_017004801.1
XP_016860290.1
25
5071
XM_017004802.1
XP_016860291.1
24
5023
XM_017004803.1
XP_016860292.1
24
4984
XM_017004804.1
XP_016860293.1
25
4939
XM_017004805.1
XP_016860294.1
24
4855
XM_017004806.1
XP_016860295.1
23
4828
XM_017004807.1
XP_016860296.1
22
4789
XM_017004808.1
XP_016860297.1
21
4564
XM_017004809.1
XP_016860298.1
29
5412
XM_017004810.1
XP_016860299.1
29
5295
XM_017004811.1
XP_016860300.1
27
5158
XM_017004812.1
XP_016860301.1
26
5108
XM_017004813.1
XP_016860302.1
12
4020
XM_017004814.1
XP_016860303.1
12
3808
XM_017004815.1
XP_016860304.1
12
3874
XM_017004816.1
XP_016860305.1
5
4183
XM_017004817.1
XP_016860306.1
10
6163
GenBank mRNA
AB209238.1
-
8
1948
AL117537.1
-
2
140
BC014042.2
-
3
644
BC116187.1
-
23
4657
40074822
BC116188.1
-
22
4630
40074825
BC126910.1
-
22
4633
BC140942.1
-
26
4864
9056754
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cell-substrate junction assembly
fibroblast migration
actin binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-2g8tf:80/10.239.165.190:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0