Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035249.1 | NP_001030326.1 | 5 | 608 | ||
GenBank mRNA | BC088226.1 | - | 5 | 526 | ||
BC105907.1 | - | 5 | 608 | |||
BC126098.1 | - | 5 | 640 | |||
BC158564.1 | - | 5 | 592 | 8374563 | ||
FQ214115.1 | - | 5 | 603 |