Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320350.1 | NP_001307279.1 | 8 | 1302 | ||
NM_001320351.1 | NP_001307280.1 | 8 | 1302 | |||
NM_003307.3 | NP_003298.1 | 8 | 1302 | |||
NR_038257.1 | 8 | 1302 | ||||
XM_005261171.3 | XP_005261228.1 | 9 | 1463 | |||
XM_011529736.2 | XP_011528038.1 | 8 | 1706 | |||
XM_017028456.1 | XP_016883945.1 | 9 | 1466 | |||
XM_017028457.1 | XP_016883946.1 | 8 | 1707 | |||
GenBank mRNA | AB001535.1 | - | 8 | 1647 | ||
AB166745.1 | - | 9 | 1415 | |||
AB166746.1 | - | 4 | 654 | |||
AJ417076.1 | - | 8 | 1215 | |||
AJ878416.1 | - | 8 | 1265 | |||
AY603182.1 | - | 8 | 1202 | |||
BC112342.1 | - | 8 | 1215 | 40069893 | ||
BI906484.1 | - | 1 | 66 |