Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prtfdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106127.1 | NP_001099597.1 | 8, 9 | 663 | ||
XM_008771860.1 | XP_008770082.1 | 9, 10 | 785 | |||
GenBank mRNA | BC166887.1 | - | 8, 9 | 660 | 6890546 | |
BC166923.1 | - | 8, 9 | 660 | 6919422 |