Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1311558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760533.1 | XP_008758755.1 | 13 | 1387 | ||
GenBank mRNA | BC085922.1 | - | 15 | 1533 | ||
EF055488.1 | - | 16 | 1543 |