Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009644.1 | NP_001009644.1 | 2 | 438 | ||
GenBank mRNA | AB197138.1 | - | 2 | 438 |