Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008890.1 | NP_001008890.1 | 1 | 48 | ||
GenBank mRNA | BC157809.1 | - | 1 | 54 | 9033183 | |
FQ221239.1 | - | 1 | 104 | |||
FQ222123.1 | - | 1 | 103 |