Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UTY SIRNA Assays ›
Gene Symbol:
UTY
Gene Name:
ubiquitously transcribed tetratricopeptide repeat containing, Y-linked
Gene Aliases:
KDM6AL, UTY1
Chromosome Location:
Chr.Y: 13230770 - 13480670 on Build GRCh38
UniGene ID:
Hs.115277
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14738
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UTY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258249.1
NP_001245178.1
17
2916
NM_001258251.1
NP_001245180.1
15
2625
NM_001258252.1
NP_001245181.1
16
2760
NM_001258253.1
NP_001245182.1
15
2625
NM_001258256.1
NP_001245185.1
16
2760
NM_001258258.1
NP_001245187.1
16
2781
NM_001258259.1
NP_001245188.1
15
2577
NM_001258260.1
NP_001245189.1
16
2715
NM_001258261.1
NP_001245190.1
16
2751
NM_001258263.1
NP_001245192.1
15
2625
NM_001258264.1
NP_001245193.1
16
2712
NM_001258265.1
NP_001245194.1
11
2376
NM_001258266.1
NP_001245195.1
16
2760
NM_001258267.1
NP_001245196.1
15
2625
NM_007125.4
NP_009056.3
15
2625
NM_182659.1
NP_872600.1
15
2625
NM_182660.1
NP_872601.1
15
2625
NR_047596.1
15
2625
NR_047597.1
16
2731
NR_047598.1
17
2861
NR_047599.1
15
2625
NR_047601.1
16
2760
NR_047602.1
14
2590
NR_047603.1
16
2748
NR_047605.1
15
2577
NR_047606.1
16
2761
NR_047607.1
17
2971
NR_047608.1
16
2712
NR_047610.1
15
2625
NR_047611.1
16
2760
NR_047612.1
17
2916
NR_047613.1
16
2866
NR_047614.1
17
2829
NR_047615.1
16
2683
NR_047616.1
16
2739
NR_047617.1
17
2887
NR_047619.1
16
2683
NR_047620.1
16
2760
NR_047621.1
17
2987
NR_047622.1
16
2817
NR_047623.1
16
2760
NR_047624.1
17
2845
NR_047625.1
14
2498
NR_047626.1
15
2625
NR_047627.1
14
2590
NR_047628.1
15
2625
NR_047629.1
15
2625
NR_047630.1
15
2625
NR_047631.1
15
2625
NR_047634.1
15
2746
NR_047635.1
15
2620
NR_047636.1
16
2715
NR_047637.1
16
2683
NR_047638.1
15
2691
NR_047639.1
14
2531
NR_047640.1
16
2731
NR_047641.1
15
2625
NR_047642.1
15
2625
NR_047643.1
16
2760
NR_047644.1
17
2818
NR_047645.1
16
2831
NR_047646.1
14
2498
NR_047647.1
15
2582
XM_005262518.3
XP_005262575.1
16
2106
XM_006724875.3
XP_006724938.1
17
2262
XM_011531441.2
XP_011529743.1
17
2262
XM_011531442.2
XP_011529744.1
17
2262
XM_011531443.2
XP_011529745.1
17
2196
XM_011531445.2
XP_011529747.1
16
2127
XM_011531446.2
XP_011529748.1
16
2106
XM_011531447.2
XP_011529749.1
16
2127
XM_011531448.2
XP_011529750.1
16
2061
XM_011531449.2
XP_011529751.1
16
2106
XM_011531450.2
XP_011529752.1
16
2061
XM_011531451.2
XP_011529753.1
15
1971
XM_011531452.2
XP_011529754.1
15
1971
XM_011531453.2
XP_011529755.1
17
2262
XM_011531454.2
XP_011529756.1
17
2262
XM_011531455.2
XP_011529757.1
17
2262
XM_011531457.2
XP_011529759.1
17
2262
XM_011531458.2
XP_011529760.1
17
2196
XM_011531459.2
XP_011529761.1
17
2262
XM_011531460.2
XP_011529762.1
17
2262
XM_011531461.2
XP_011529763.1
17
2262
XM_011531462.2
XP_011529764.1
17
2196
XM_011531463.2
XP_011529765.1
16
2106
XM_011531464.2
XP_011529766.1
16
2106
XM_011531465.2
XP_011529767.1
15
1971
XM_017030066.1
XP_016885555.1
17
2196
XM_017030067.1
XP_016885556.1
16
2106
XM_017030068.1
XP_016885557.1
15
1971
XM_017030069.1
XP_016885558.1
15
1971
XM_017030070.1
XP_016885559.1
17
2262
XM_017030071.1
XP_016885560.1
17
2262
XM_017030072.1
XP_016885561.1
16
2061
XM_017030073.1
XP_016885562.1
15
1971
XM_017030074.1
XP_016885563.1
17
2196
GenBank mRNA
AF000994.1
-
15
2625
AF000995.1
-
15
2625
AF000996.1
-
15
2625
AK292858.1
-
15
1875
AK308952.1
-
1
224
AY682463.1
-
14
1527
AY730613.1
-
16
1776
AY769061.1
-
17
1856
AY769063.1
-
15
1651
AY769064.1
-
14
1585
AY769065.1
-
15
1620
AY769066.1
-
17
1911
AY769067.1
-
15
1620
AY769068.1
-
16
1678
AY769069.1
-
14
1585
AY934848.1
-
15
1620
AY934849.1
-
16
1726
AY934851.1
-
15
1572
AY934852.1
-
15
1611
AY934853.1
-
15
1601
AY934854.1
-
14
1521
AY934855.1
-
14
1521
AY934856.1
-
14
1537
AY934857.1
-
16
1734
CR936684.1
-
7
723
DQ140389.1
-
16
1755
DQ341385.1
-
15
1620
DQ341386.1
-
15
1620
DQ341387.1
-
14
1521
DQ345542.1
-
16
1810
DQ345543.1
-
16
1710
DQ350457.1
-
15
1620
DQ350458.1
-
16
1734
DQ350459.1
-
15
1620
EF612661.1
-
14
1521
EF612663.1
-
16
1755
EF612665.1
-
15
1572
EF612667.1
-
14
1521
EF612668.1
-
17
1911
EF612670.1
-
13
1428
EF612672.1
-
15
1572
EF612673.1
-
16
1776
EF612674.1
-
15
1572
EF612675.1
-
16
1707
EF612677.1
-
17
1911
EF612680.1
-
16
1710
EF612683.1
-
16
1746
EF612685.1
-
17
1845
EF612686.1
-
16
1755
EF612687.1
-
15
1668
EF612689.1
-
16
1755
EF612690.1
-
14
1521
EF612691.1
-
16
1755
EF612692.1
-
13
1530
EF612693.1
-
15
1620
EF612695.1
-
16
1707
EF612696.1
-
16
1710
EF612697.1
-
15
1620
EF612698.1
-
13
1530
EF653165.1
-
14
1585
EF653166.1
-
14
1526
EF653168.1
-
16
1755
EF653169.1
-
14
1585
EF653170.1
-
13
1486
EF653171.1
-
15
1620
EF653172.1
-
14
1511
EF653173.1
-
15
1620
EF653174.1
-
17
1816
EF653175.1
-
17
1861
EF653176.1
-
14
1570
EF653178.1
-
18
1977
EF653179.1
-
15
1620
EF653180.1
-
16
1726
EF653181.1
-
16
1710
EF653182.1
-
14
1585
EF653183.1
-
16
1726
EF653184.1
-
16
1734
EF653186.1
-
16
1743
EF653189.1
-
11
1371
EF653190.1
-
16
1755
EF653192.1
-
14
1526
EF653193.1
-
16
1726
EF653194.1
-
16
1700
EF653195.1
-
16
1726
EF653196.1
-
15
1620
EF653197.1
-
14
1585
EF653198.1
-
15
1620
EF653199.1
-
17
1840
EF653200.1
-
16
1755
EF653201.1
-
15
1661
EF653202.1
-
13
1399
EF653203.1
-
14
1585
EF653204.1
-
16
1736
EF653207.1
-
17
1813
EF653208.1
-
16
1726
EF653209.1
-
16
1826
EF653211.1
-
17
1784
EF653212.1
-
15
1620
EF653213.1
-
14
1493
EF653214.1
-
17
1937
EF653215.1
-
15
1690
EF653216.1
-
15
1672
EF653217.1
-
15
1577
EF653218.1
-
15
1572
EF653219.1
-
17
1911
EF653221.1
-
13
1486
EF653223.1
-
16
1700
EF653224.1
-
17
1966
EF685283.1
-
16
1756
EF685285.1
-
16
1707
EF685286.1
-
16
1755
EF685287.1
-
15
1620
EF685288.1
-
17
1911
EF685289.1
-
8
1007
EF685290.1
-
16
1861
EF685291.1
-
17
1824
EF685292.1
-
16
1678
EF685293.1
-
16
1734
EF685294.1
-
17
1882
EF685296.1
-
16
1678
EF685297.1
-
16
1755
EF685298.1
-
17
1982
EF685299.1
-
16
1726
EF685300.1
-
16
1812
EF685301.1
-
16
1755
EF685302.1
-
15
1593
EF685303.1
-
15
1620
EF685304.1
-
15
1643
EF685305.1
-
16
1695
EF685306.1
-
15
1656
EF685307.1
-
17
1840
EF685308.1
-
14
1493
EF685309.1
-
15
1620
EF685310.1
-
15
1620
EF685311.1
-
15
1620
EF685312.1
-
14
1585
EF685313.1
-
14
1585
EF685314.1
-
15
1620
EF685315.1
-
14
1526
EF685316.1
-
15
1620
EF685317.1
-
16
1776
EF685318.1
-
14
1521
EF685319.1
-
14
1521
EF685320.1
-
15
1572
EF685321.1
-
15
1620
EF685322.1
-
14
1521
EF685323.1
-
14
1521
EF685324.1
-
14
1437
EF685325.1
-
15
1620
EF685326.1
-
16
1678
EF685327.1
-
16
1695
EF685328.1
-
14
1521
EF685329.1
-
13
1530
EF685330.1
-
16
1812
EF685331.1
-
1620
EF685332.1
-
16
1755
EF685333.1
-
17
1746
EF685334.1
-
15
1620
EF685335.1
-
11
1304
EF685338.1
-
15
1450
EF685339.1
-
14
1521
EF685340.1
-
14
1521
EF685341.1
-
14
1521
EF685342.1
-
15
1620
EU026277.1
-
16
1686
EU026279.1
-
13
1482
EU026280.1
-
17
1836
EU026281.1
-
14
1585
EU026282.1
-
13
1427
EU026284.1
-
15
1616
EU026286.1
-
14
1537
EU026287.1
-
13
1399
EU026289.1
-
14
1527
EU026290.1
-
15
1821
EU026294.1
-
14
1585
EU026297.1
-
16
1722
EU026298.1
-
15
1651
EU026299.1
-
14
1585
EU026300.1
-
14
1585
EU026301.1
-
16
1757
EU026302.1
-
17
1884
EU026303.1
-
16
1750
EU026305.1
-
14
1521
EU026306.1
-
16
1734
EU026307.1
-
15
1620
EU026308.1
-
14
1592
EU026309.1
-
17
1797
EU026310.1
-
15
1620
EU026311.1
-
16
1755
EU026313.1
-
15
1572
EU026314.1
-
17
1861
EU026315.1
-
15
1651
EU026318.1
-
13
1487
EU026319.1
-
16
1678
EU026320.1
-
18
1919
EU026322.1
-
15
1615
EU026323.1
-
15
1715
EU026324.1
-
13
1410
EU026326.1
-
16
1726
EU026327.1
-
15
1691
EU026328.1
-
15
1675
EU026330.1
-
13
1525
EU026331.1
-
15
1620
EU026332.1
-
13
1486
EU026333.1
-
16
1784
EU026334.1
-
14
1518
EU026335.1
-
17
1813
EU026336.1
-
10
1251
EU026337.1
-
12
1359
EU026338.1
-
15
1620
EU026339.1
-
13
1486
EU026340.1
-
15
1706
EU026341.1
-
8
1019
EU026342.1
-
15
1728
EU026343.1
-
16
1742
EU026344.1
-
14
1537
EU026345.1
-
15
1615
EU026346.1
-
13
1471
EU026347.1
-
13
1486
EU026348.1
-
15
1620
EU026349.1
-
15
1620
EU026351.1
-
15
1620
EU116343.1
-
10
1148
EU116346.1
-
15
1620
EU116348.1
-
15
1621
EU116349.1
-
15
1714
EU116350.1
-
15
1548
EU116351.1
-
9
1084
EU116352.1
-
15
1615
EU116353.1
-
16
1710
EU163941.1
-
15
1686
EU163942.1
-
16
1678
EU163943.1
-
6
843
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of gene expression
oxidation-reduction process
histone H3-K27 demethylation
histone demethylase activity
metal ion binding
dioxygenase activity
histone demethylase activity (H3-K27 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-2g8tf:80/10.239.165.190:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0