Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam57b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106296.2 | NP_001099766.2 | 2 | 358 | ||
XM_006230237.2 | XP_006230299.1 | 3 | 434 | |||
XM_006230239.2 | XP_006230301.1 | 2 | 179 | |||
XM_006230240.2 | XP_006230302.1 | 2 | 954 | |||
GenBank mRNA | BC161920.1 | - | 2 | 358 | 8370175 |