Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005663.4 | NP_005654.3 | 2 | 598 | ||
XM_017008589.1 | XP_016864078.1 | 3 | 1419 | |||
GenBank mRNA | AB044549.1 | - | 1 | 385 | ||
AF101434.1 | - | 2 | 327 | |||
AK001304.1 | - | 2 | 385 | |||
AK095737.1 | - | 1 | 641 | |||
AK126056.1 | - | 2 | 1473 | |||
AK222853.1 | - | 2 | 335 | |||
BC002764.2 | - | 2 | 338 | 3630594 |