Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100836.1 | NP_001094306.1 | 6 | 913 | ||
GenBank mRNA | BC158809.1 | - | 6 | 914 | 7364883 | |
CF113031.1 | - | 3 | 511 |