Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047862.1 | NP_001041327.1 | 5 | 513 | ||
XM_008761741.1 | XP_008759963.1 | 4 | 541 | |||
GenBank mRNA | AY325167.1 | - | 5 | 513 | ||
FQ226210.1 | - | 3 | 375 | |||
FQ230936.1 | - | 3 | 375 | |||
FQ234165.1 | - | 3 | 375 |