Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106603.1 | NP_001100073.1 | 11 | 1920 | ||
GenBank mRNA | FQ214319.1 | - | 11 | 1290 |