Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106609.2 | NP_001100079.2 | 14 | 1863 | ||
NM_001184712.1 | NP_001171641.1 | 14 | 1863 | |||
GenBank mRNA | BF564749.1 | - | 2 | 156 | ||
CK603679.1 | - | 6 | 559 |