Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025409.1 | NP_001020580.1 | 5 | 443 | ||
GenBank mRNA | BC059156.1 | - | 5 | 460 | ||
BC097943.1 | - | 5 | 443 | 7458532 | ||
BC107476.1 | - | 5 | 440 | 7443793 |