Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF75A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302109.1 | NP_001289038.1 | 7 | 1603 | ||
NM_001324041.1 | NP_001310970.1 | 7 | 1560 | |||
NM_001324085.1 | NP_001311014.1 | 5 | 1296 | |||
NM_153028.3 | NP_694573.1 | 6 | 1079 | |||
NR_126069.1 | 5 | 1238 | ||||
NR_136681.1 | 6 | 1407 | ||||
NR_136683.1 | 5 | 883 | ||||
NR_136686.1 | 6 | 1433 | ||||
XM_011522646.2 | XP_011520948.1 | 6 | 1492 | |||
XM_011522647.2 | XP_011520949.1 | 6 | 1177 | |||
XM_017023642.1 | XP_016879131.1 | 6 | 975 | |||
GenBank mRNA | AK056091.1 | - | 6 | 1002 | ||
AK093979.1 | - | 5 | 1191 | |||
BC119652.1 | - | 5 | 804 | 40115880 | ||
BC119653.1 | - | 5 | 804 | 40115881 | ||
CR749782.1 | - | 6 | 902 |