Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Theg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013102.1 | NP_001013120.1 | 1 | 290 | ||
XM_006240857.2 | XP_006240919.1 | 1 | 310 | |||
XM_006240858.2 | XP_006240920.1 | 1 | 310 | |||
GenBank mRNA | BC083921.1 | - | 1 | 290 | 7135133 |