Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099474.1 | NP_001092944.1 | 3 | 554 | ||
XM_008765199.1 | XP_008763421.1 | 4 | 563 |
Gene Symbol: LOC103690080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765206.1 | XP_008763428.1 | 3 | 545 | ||
XM_008765207.1 | XP_008763429.1 | 3 | 545 |
Gene Symbol: Vom2r56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099484.1 | NP_001092954.1 | 554 |
Set Membership: |
G-protein coupled receptor |