Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011989.1 | NP_001011989.1 | 7 | 844 | ||
XM_006241396.2 | XP_006241458.1 | 7 | 940 | |||
GenBank mRNA | BC087741.1 | - | 7 | 844 | 7314130 |
Gene Symbol: LOC100909505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750325.3 | XP_003750373.1 | 941 |