Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC300308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013952.1 | NP_001013974.1 | 1084 | |||
GenBank mRNA | BC090074.1 | - | 1084 | 7114534 |
Gene Symbol: RGD1566251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765268.1 | XP_008763490.1 | 2 | 4558 | ||
XM_008765269.1 | XP_008763491.1 | 2 | 4558 | |||
XM_008765270.1 | XP_008763492.1 | 3 | 4420 | |||
XM_008765271.1 | XP_008763493.1 | 2 | 4558 | |||
XM_008765272.1 | XP_008763494.1 | 2 | 4558 | |||
XM_008765273.1 | XP_008763495.1 | 2 | 4558 | |||
XM_008765274.1 | XP_008763496.1 | 3 | 4739 | |||
XM_008765275.1 | XP_008763497.1 | 2 | 4558 | |||
XM_008765276.1 | XP_008763498.1 | 2 | 4544 | |||
XM_008765277.1 | XP_008763499.1 | 2 | 4558 | |||
XM_008765278.1 | XP_008763500.1 | 3 | 4455 | |||
XM_008765279.1 | XP_008763501.1 | 2 | 4558 | |||
XM_008765280.1 | XP_008763502.1 | 2 | 4558 | |||
XM_008765281.1 | XP_008763503.1 | 2 | 4558 | |||
XM_008765282.1 | XP_008763504.1 | 2 | 4558 | |||
XM_008765283.1 | XP_008763505.1 | 2 | 4558 | |||
XM_008765284.1 | XP_008763506.1 | 2 | 4558 | |||
XM_008765285.1 | XP_008763507.1 | 2 | 4558 |