Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009670.1 | NP_001009670.1 | 5 | 975 | ||
XM_006243717.2 | XP_006243779.1 | 4 | 816 | |||
XM_006243720.2 | XP_006243782.1 | 4 | 1570 | |||
XM_008766494.1 | XP_008764716.1 | 5 | 1387 | |||
XM_008766495.1 | XP_008764717.1 | 6 | 1041 | |||
XM_008766496.1 | XP_008764718.1 | 5 | 990 | |||
XM_008766497.1 | XP_008764719.1 | 5 | 859 | |||
XM_008766498.1 | XP_008764720.1 | 4 | 811 | |||
GenBank mRNA | BC088472.1 | - | 5 | 975 | 7128506 |