Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107082.1 | NP_001100552.1 | 6 | 464 | ||
XM_008768798.1 | XP_008767020.1 | 6 | 477 |