Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177421.3 | NP_803156.2 | 8, 9 | 1615 | ||
XM_006251965.1 | XP_006252027.1 | 8, 9 | 2253 | |||
GenBank mRNA | AB040056.1 | - | 8, 9 | 1622 | ||
BC099072.1 | - | 8, 9 | 1615 | 7312276 | ||
FQ212273.1 | - | 8, 9 | 1617 |